Gene Transcripts
- Gene Symbol
- TRGV3
- Entrez Gene ID
- 6976
- Gene Name
- T cell receptor gamma variable 3
- Gene Aliases
- TCRGV3, V1S3
- Chromosome Location
- Chr. 7: 38358512 - 38358979 on Build GRCh38
- UniGene ID
- Hs.731375
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC071739.1 |
- |
2 |
901 |
|
View Full Product Details