Gene Transcripts

Gene Symbol
RAB40AL
Entrez Gene ID
282808
Gene Name
RAB40A, member RAS oncogene family-like
Gene Aliases
MRXSMP, RAR2, RLGP
Chromosome Location
Chr. X: 102937272 - 102938300 on Build GRCh38
UniGene ID
Hs.449517
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001031834.1 NP_001027004.1 1 930
GenBank mRNA BC101169.1 - 1 930
BC101170.2 - 1 930 40016065
BC101171.1 - 1 930 40016067
BC101172.1 - 1 930 40016069

View Full Product Details