Gene Transcripts

Gene Symbol
Vom1r48
Entrez Gene ID
494279
Gene Name
vomeronasal 1 receptor 48
Gene Aliases
V1rg13
Chromosome Location
Chr. 1: 66972358 - 66973219 on Build Rnor_6.0
UniGene ID
Rn.129419
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008931.1 NP_001008931.1 198
GenBank mRNA AY510315.1 - 198

Gene Symbol
LOC103691006
Entrez Gene ID
103691006
Gene Name
vomeronasal type-1 receptor 4-like
Gene Aliases
Chromosome Location
Chr. 1: 66959647 - 66960933 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008758859.1 XP_008757081.1 1 215

View Full Product Details