Gene Transcripts

Gene Symbol
Spata24
Entrez Gene ID
291676
Gene Name
spermatogenesis associated 24
Gene Aliases
RGD1311742
Chromosome Location
Chr. 18: 28446592 - 28454725 on Build Rnor_6.0
UniGene ID
Rn.136857
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025636.2 NP_001020807.2 4 539
GenBank mRNA DQ080016.1 - 4 539

View Full Product Details