Gene Transcripts

Gene Symbol
Cdr2
Entrez Gene ID
308958
Gene Name
cerebellar degeneration-related protein 2
Gene Aliases
RGD1310578
Chromosome Location
Chr. 1: 190889698 - 190914606 on Build Rnor_6.0
UniGene ID
Rn.3537
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025682.1 NP_001020853.1 3 354
GenBank mRNA BC098028.1 - 3 354

View Full Product Details