Gene Transcripts

Gene Symbol
Ydjc
Entrez Gene ID
69101
Gene Name
YdjC homolog (bacterial)
Gene Aliases
1810015A11Rik, 4930521M19Rik
Chromosome Location
Chr. 16: 17146882 - 17148857 on Build GRCm38
UniGene ID
Mm.101656
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026940.4 NP_081216.1 2 283
GenBank mRNA AK007503.1 - 2 272
AK032396.1 - 313
AK076869.1 - 894
AK140044.1 - 388
AK141482.1 - 1 358
BC030425.1 - 2 285
BC115584.1 - 2 271 40102346
BC115585.1 - 2 271 40102347

View Full Product Details