Gene Transcripts

Gene Symbol
Gpr17
Entrez Gene ID
574402
Gene Name
G protein-coupled receptor 17
Gene Aliases
AI853548
Chromosome Location
Chr. 18: 31942999 - 31949636 on Build GRCm38
UniGene ID
Mm.391108
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025381.2 NP_001020552.1 2 1292
GenBank mRNA BC070439.1 - 2 1292

View Full Product Details