Gene Transcripts

Gene Symbol
OR5K2
Entrez Gene ID
402135
Gene Name
olfactory receptor family 5 subfamily K member 2
Gene Aliases
OR3-9
Chromosome Location
Chr. 3: 98497681 - 98498631 on Build GRCh38
UniGene ID
Hs.632558
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004737.1 NP_001004737.1 1 581
GenBank mRNA BC136724.1 - 1 658
BC136725.1 - 1 658 9020715

View Full Product Details