Gene Transcripts

Gene Symbol
PQBP1
Entrez Gene ID
10084
Gene Name
polyglutamine binding protein 1
Gene Aliases
MRX2, MRX55, MRXS3, MRXS8, NPW38, RENS1, SHS
Chromosome Location
Chr. X: 48897862 - 48903145 on Build GRCh38
UniGene ID
Hs.534384
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001032381.1 NP_001027553.1 4, 5 421
NM_001032382.1 NP_001027554.1 4, 5 489
NM_001032383.1 NP_001027555.1 4, 5 500
NM_001032384.1 NP_001027556.1 4, 5 409
NM_001167989.1 NP_001161461.1 4, 5 421
NM_001167990.1 NP_001161462.1 4, 5 379
NM_005710.2 NP_005701.1 3, 4 554
XM_005272571.3 XP_005272628.1 4, 5 484
XM_011543884.2 XP_011542186.1 4, 5 559
XM_017029207.1 XP_016884696.1 4, 5 550
GenBank mRNA AB016533.1 - 4, 5 470
AB041833.1 - 4 314
AB041834.1 - 3, 4 300
AB041835.1 - 3, 4 300
AJ005893.1 - 3, 4 557
AJ242829.1 - 4, 5 360
AJ973594.1 - 3, 4 315
AJ973595.1 - 3, 4 315
AJ973596.1 - 315
AJ973597.1 - 3, 4 315
AJ973598.1 - 3, 4 315
AJ973599.1 - 3, 4 315
AJ973600.1 - 3, 4 315
AJ973601.1 - 4, 5 402
AJ973602.1 - 3, 4 619
AJ973603.1 - 4, 5 402
AJ973604.1 - 3, 4 319
AJ973606.1 - 3, 4 315
BC012358.1 - 4, 5 381 4662649
BE385548.1 - 2, 3 235
BE396796.1 - 4, 5 401
BI818141.1 - 4, 5 383
BX362311.2 - 4, 5 458
BX369697.2 - 4, 5 641
CD051018.1 - 4, 5 400
CN292761.1 - 4, 5 489
CN292769.1 - 4, 5 407
KU178500.1 - 3, 4 300
KU178501.1 - 4, 5 348

View Full Product Details