Gene Transcripts

Gene Symbol
UQCRH
Entrez Gene ID
7388
Gene Name
ubiquinol-cytochrome c reductase hinge protein
Gene Aliases
QCR6, UQCR8
Chromosome Location
Chr. 1: 46303613 - 46316777 on Build GRCh38
UniGene ID
Hs.481571
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001297565.1 NP_001284494.1 5 697
NM_001297566.1 NP_001284495.1 3 522
NM_006004.3 NP_005995.2 4 549
GenBank mRNA BC001426.2 - 4 449
BC001934.1 - 4 454 2823237
BC015177.1 - 4 452 3905169
BC093060.1 - 5 612 30409772
BC107703.1 - 4 455 6605146
BG472687.1 - 3 423

View Full Product Details