Gene Transcripts

Gene Symbol
OVOL2
Entrez Gene ID
58495
Gene Name
ovo like zinc finger 2
Gene Aliases
EUROIMAGE566589, PPCD1, ZNF339
Chromosome Location
Chr. 20: 18024152 - 18059188 on Build GRCh38
UniGene ID
Hs.661013
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001303461.1 NP_001290390.1 4 1348
NM_001303462.1 NP_001290391.1 3 937
NM_021220.3 NP_067043.2 4 1397
GenBank mRNA AK022284.1 - 4 1392
AL079276.1 - 3 976
BC006148.1 - 4 1396 3959436

View Full Product Details