Gene Transcripts

Gene Symbol
GPATCH3
Entrez Gene ID
63906
Gene Name
G-patch domain containing 3
Gene Aliases
GPATC3
Chromosome Location
Chr. 1: 26890488 - 26900471 on Build GRCh38
UniGene ID
Hs.10903
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022078.2 NP_071361.2 3, 4 1085
GenBank mRNA AK022517.1 - 3, 4 1080
AK057930.1 - 3, 4 1080
AK303183.1 - 4, 5 1028
AK308100.1 - 3, 4 948
AL833886.1 - 3, 4 1067
BC007767.2 - 3, 4 1061 4298740
CR936767.1 - 3, 4 1085

View Full Product Details