Gene Transcripts

Gene Symbol
PKN1
Entrez Gene ID
5585
Gene Name
protein kinase N1
Gene Aliases
DBK, PAK-1, PAK1, PKN, PKN-ALPHA, PRK1, PRKCL1
Chromosome Location
Chr. 19: 14433354 - 14471867 on Build GRCh38
UniGene ID
Hs.466044
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002741.3 NP_002732.3 4 689
NM_213560.1 NP_998725.1 4 556
XM_011528128.1 XP_011526430.1 4 572
GenBank mRNA AK123007.1 - 4 570
AK292130.1 - 4 553
AK313886.1 - 4 529
BC040061.1 - 4 686 5752583
BC094766.1 - 4 720 30342125
D26181.1 - 4 557
S75546.1 - 4 605
U33053.1 - 4 605

View Full Product Details