Gene Transcripts

Gene Symbol
NAA40
Entrez Gene ID
79829
Gene Name
N(alpha)-acetyltransferase 40, NatD catalytic subunit
Gene Aliases
NAT11, PATT1
Chromosome Location
Chr. 11: 63938970 - 63957322 on Build GRCh38
UniGene ID
Hs.523753
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001300800.1 NP_001287729.1 8 2775
NM_024771.3 NP_079047.2 8 2704
XM_006718689.3 XP_006718752.1 8 2601
XM_011545254.2 XP_011543556.1 9 2778
XM_011545255.2 XP_011543557.1 9 2688
GenBank mRNA AK023910.1 - 8 2673
BC024160.2 - 1 1961
BC041617.1 - 8 2633 5921480
BC052298.1 - 8 2633 6292760
BX647309.1 - 7 3729
JF263566.1 - 8 2673

View Full Product Details