Gene Transcripts

Gene Symbol
TRIM45
Entrez Gene ID
80263
Gene Name
tripartite motif containing 45
Gene Aliases
RNF99
Chromosome Location
Chr. 1: 117107711 - 117121789 on Build GRCh38
UniGene ID
Hs.301526
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145635.1 NP_001139107.1 1 833
NM_025188.3 NP_079464.2 1 833
GenBank mRNA AK023243.1 - 1 772
AK222901.1 - 1 777
AY669488.1 - 1 833
BC034943.2 - 1 806 4826874

View Full Product Details