Gene Transcripts

Gene Symbol
PUS1
Entrez Gene ID
80324
Gene Name
pseudouridylate synthase 1
Gene Aliases
MLASA1
Chromosome Location
Chr. 12: 131929200 - 131944040 on Build GRCh38
UniGene ID
Hs.592004
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002019.2 NP_001002019.1 3 482
NM_001002020.2 NP_001002020.1 3 511
NM_025215.5 NP_079491.2 3 859
XM_011538768.2 XP_011537070.1 2 124
XM_011538769.2 XP_011537071.1 3 859
GenBank mRNA AF116238.1 - 2 219
AF318369.1 - 3 851
AK074659.1 - 3 859
AK292242.1 - 3 455
BC002901.2 - 3 425 3943243
BC009505.2 - 3 630 2822709
BC019320.1 - 3 630 2822709
BC035964.1 - 2 999
BM923133.1 - 3 488
BX340929.2 - 3 484
CB118952.1 - 3 511

View Full Product Details