Gene Transcripts

Gene Symbol
LIMD2
Entrez Gene ID
80774
Gene Name
LIM domain containing 2
Gene Aliases
Chromosome Location
Chr. 17: 63695889 - 63701222 on Build GRCh38
UniGene ID
Hs.591166
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030576.3 NP_085053.1 5 1909
GenBank mRNA AK092301.1 - 5 1875

View Full Product Details