Gene Transcripts

Gene Symbol
TLR10
Entrez Gene ID
81793
Gene Name
toll like receptor 10
Gene Aliases
CD290
Chromosome Location
Chr. 4: 38772238 - 38782990 on Build GRCh38
UniGene ID
Hs.120551
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017388.2 NP_001017388.1 2 492
NM_001195106.1 NP_001182035.1 3 809
NM_001195107.1 NP_001182036.1 3 681
NM_001195108.1 NP_001182037.1 3 712
NM_030956.3 NP_112218.2 4 998
XM_011513760.2 XP_011512062.1 2 386
XM_011513761.2 XP_011512063.1 5 1079
XM_011513762.2 XP_011512064.1 4 759
GenBank mRNA AB445680.1 - 1 360
AF296673.1 - 3 846
AK292026.1 - 2 478
AK313468.1 - 2 629
AY358300.1 - 2 470
BC089406.1 - 2 844 3066588
BC109111.2 - 1 967 40035401
BC109112.2 - 2 510 40035404
DA643313.1 - 2 492
DQ011502.1 - 1 360
DQ011503.1 - 1 360
DQ011504.1 - 1 360
DQ011505.1 - 1 360
DQ011506.1 - 1 360
DQ011507.1 - 1 360
DQ011508.1 - 1 360
DQ011509.1 - 1 360
DQ011510.1 - 1 360
DQ011511.1 - 1 360
DQ011512.1 - 1 360
DQ011513.1 - 1 360
DQ011514.1 - 1 360
DQ011515.1 - 1 360
DQ011516.1 - 1 360

View Full Product Details