Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_032214.3 | NP_115590.1 | 4 | 660 | |
NM_175077.2 | NP_778252.1 | 4 | 660 | ||
XM_017028098.1 | XP_016883587.1 | 4 | 651 | ||
GenBank mRNA | AF290985.1 | - | 3 | 237 | |
AF290986.1 | - | 3 | 237 | ||
AF326353.1 | - | 3 | 237 | ||
AK025645.1 | - | 4 | 297 | ||
AK291513.1 | - | 4 | 640 | ||
BC042041.1 | - | 4 | 599 | 4429896 | |
BQ052468.1 | - | 4 | 325 |