Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_033125.3 | NP_149116.2 | 2 | 241 | |
XM_011536204.2 | XP_011534506.1 | 2 | 255 | ||
XM_011536205.2 | XP_011534507.1 | 2 | 213 | ||
XM_011536210.1 | XP_011534512.1 | 2 | 213 | ||
GenBank mRNA | AF268892.1 | - | 2 | 174 | |
AK304546.1 | - | 2 | 213 | ||
AY145502.1 | - | 2 | 174 | ||
BC012014.1 | - | 2 | 172 | ||
BC037247.1 | - | 2 | 189 | ||
BC047565.1 | - | 2 | 241 | 4838011 | |
GQ472228.1 | - | 2 | 241 |