Gene Transcripts

Gene Symbol
MRFAP1
Entrez Gene ID
93621
Gene Name
Morf4 family associated protein 1
Gene Aliases
PAM14, PGR1
Chromosome Location
Chr. 4: 6640091 - 6642745 on Build GRCh38
UniGene ID
Hs.406590
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001272053.1 NP_001258982.1 3 1361
NM_001272054.1 NP_001258983.1 3 1408
NM_033296.2 NP_150638.1 2 1480
GenBank mRNA AK057736.1 - 3 834
AK074496.1 - 2 887
AK123360.1 - 3 1361
AK129651.1 - 3 765
AK300242.1 - 3 826
AK303974.1 - 3 807
BC022797.1 - 2 866 5434657
BI858568.1 - 1 28
CB052459.1 - 1 247

View Full Product Details