Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_080282.3 | NP_525021.3 | 19 | 2941 | |
GenBank mRNA | AL832004.2 | - | 19 | 2647 | |
AY028900.1 | - | 19 | 2941 | ||
AY247065.1 | - | 19 | 2972 | ||
BC136460.1 | - | 16 | 2146 | ||
BC144124.1 | - | 16 | 2215 |