Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001144884.1 | NP_001138356.1 | 5 | 682 | |
NM_133496.4 | NP_598003.2 | 5 | 680 | ||
XM_011540779.2 | XP_011539081.1 | 6 | 779 | ||
XM_017000400.1 | XP_016855889.1 | 5 | 680 | ||
XM_017000401.1 | XP_016855890.1 | 5 | 680 | ||
GenBank mRNA | AF233345.1 | - | 5 | 510 | |
AF529197.1 | - | 5 | 495 | ||
AK313638.1 | - | 5 | 663 | ||
AY094606.1 | - | 5 | 495 | ||
BC064692.1 | - | 5 | 636 | 30330665 | |
BX537375.1 | - | 5 | 682 | ||
CR936806.1 | - | 5 | 676 |