Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_144579.2 | NP_653180.1 | 11 | 814 | |
XM_005264648.1 | XP_005264705.1 | 11 | 697 | ||
XM_011533186.2 | XP_011531488.1 | 12 | 2362 | ||
XM_011533187.2 | XP_011531489.1 | 12 | 2359 | ||
XM_011533188.2 | XP_011531490.1 | 11 | 2217 | ||
XM_011533189.1 | XP_011531491.1 | 11 | 720 | ||
XM_011533190.2 | XP_011531492.1 | 12 | 1267 | ||
XM_011533192.2 | XP_011531494.1 | 12 | 1501 | ||
XM_011533194.2 | XP_011531496.1 | 12 | 2360 | ||
XM_011533196.2 | XP_011531498.1 | 11 | 1446 | ||
XM_011533197.1 | XP_011531499.1 | 11 | 697 | ||
XM_017005340.1 | XP_016860829.1 | 11 | 2214 | ||
XM_017005341.1 | XP_016860830.1 | 11 | 2215 | ||
XM_017005342.1 | XP_016860831.1 | 11 | 720 | ||
XM_017005344.1 | XP_016860833.1 | 11 | 1446 | ||
XM_017005345.1 | XP_016860834.1 | 3 | 1819 | ||
XM_017005346.1 | XP_016860835.1 | 11 | 664 | ||
GenBank mRNA | AK289731.1 | - | 11 | 703 | |
AK294751.1 | - | 11 | 751 | ||
AK295637.1 | - | 11 | 602 | ||
AK307116.1 | - | 2 | 167 | ||
AK308863.1 | - | 2 | 154 | ||
AK309091.1 | - | 2 | 284 | ||
AK315861.1 | - | 1 | 40 | ||
AY044437.1 | - | 11 | 818 | ||
BC101311.2 | - | 11 | 814 | 40024811 | |
BC101312.1 | - | 11 | 814 | 40024812 | |
BC101313.2 | - | 11 | 814 | 40024813 |