Gene Transcripts
- Gene Symbol
- FAM210A
- Entrez Gene ID
- 125228
- Gene Name
- family with sequence similarity 210 member A
- Gene Aliases
- C18orf19, HsT2329
- Chromosome Location
- Chr. 18: 13663347 - 13726592 on Build GRCh38
- UniGene ID
- Hs.13034
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK055618.1 |
- |
3 |
544 |
|
View Full Product Details