Gene Transcripts

Gene Symbol
YTHDF3
Entrez Gene ID
253943
Gene Name
YTH N6-methyladenosine RNA binding protein 3
Gene Aliases
Chromosome Location
Chr. 8: 63168553 - 63212788 on Build GRCh38
UniGene ID
Hs.491861
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001277813.1 NP_001264742.1 6 2338
NM_001277814.1 NP_001264743.1 6 2092
NM_001277815.1 NP_001264744.1 6 2223
NM_001277816.1 NP_001264745.1 4 2106
NM_001277817.1 NP_001264746.1 5 2031
NM_001277818.1 NP_001264747.1 5 1952
NM_152758.5 NP_689971.4 5 2192
NR_102434.1 4 593
XM_011517509.2 XP_011515811.1 6 2185
XM_011517510.2 XP_011515812.1 4 1948
GenBank mRNA AK056219.1 - 6 1976
AK093081.1 - 5 1952
AK127574.1 - 5 2124
AL831816.1 - 5 2027
AL832005.1 - 5 2125
AL832335.1 - 6 2120
AL832369.1 - 3 1826
BC052970.1 - 5 2141 6044054
BX537996.2 - 5 2129
CN335163.1 - 4 580

View Full Product Details