Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_153486.3 | NP_705690.2 | 9 | 1278 | |
NM_194436.2 | NP_919417.1 | 9 | 1209 | ||
GenBank mRNA | AY092767.1 | - | 9 | 1214 | |
BC040279.1 | - | 9 | 1243 | 5162826 | |
BC047902.1 | - | 9 | 1273 | 5762691 |