Gene Transcripts
- Gene Symbol
- CDC7
- Entrez Gene ID
- 8317
- Gene Name
- cell division cycle 7
- Gene Aliases
- CDC7L1, HsCDC7, Hsk1, huCDC7
- Chromosome Location
- Chr. 1: 91500814 - 91525764 on Build GRCh38
- UniGene ID
- Hs.533573
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001134419.1 |
NP_001127891.1
|
2, 3 |
254 |
|
NM_001134420.1 |
NP_001127892.1
|
2, 3 |
382 |
|
NM_003503.3 |
NP_003494.1
|
2, 3 |
288 |
|
XM_005271241.2 |
XP_005271298.1
|
2, 3 |
250 |
|
XM_005271244.2 |
XP_005271301.1
|
2, 3 |
288 |
|
XM_017002425.1 |
XP_016857914.1
|
2, 3 |
250 |
|
XM_017002426.1 |
XP_016857915.1
|
2, 3 |
233 |
|
GenBank mRNA |
AB003698.1 |
- |
2, 3 |
255 |
|
AB209337.1 |
- |
1, 2 |
964 |
|
AF005209.1 |
- |
1, 2 |
123 |
|
AF015592.1 |
- |
2, 3 |
268 |
|
AK312726.1 |
- |
2, 3 |
344 |
|
BC110526.1 |
- |
2, 3 |
211 |
40034178 |
BC110527.1 |
- |
2, 3 |
245 |
40034179 |
BC111044.1 |
- |
2, 3 |
268 |
5518616 |
DA127303.1 |
- |
2, 3 |
382 |
|
DA791490.1 |
- |
2, 3 |
288 |
|
View Full Product Details