Gene Transcripts

Gene Symbol
NDUFC1
Entrez Gene ID
4717
Gene Name
NADH:ubiquinone oxidoreductase subunit C1
Gene Aliases
KFYI
Chromosome Location
Chr. 4: 139289917 - 139302551 on Build GRCh38
UniGene ID
Hs.84549
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001184986.1 NP_001171915.1 4 911
NM_001184987.1 NP_001171916.1 3 379
NM_001184988.1 NP_001171917.1 4 415
NM_001184989.1 NP_001171918.1 4 438
NM_001184990.1 NP_001171919.1 3 375
NM_001184991.1 NP_001171920.1 3 852
NM_002494.3 NP_002485.1 3 514
GenBank mRNA AF047184.1 - 2 109
AF047435.1 - 2 89
AK291147.1 - 2 119
BC101798.1 - 2 125 8069304
BC107682.1 - 3 484 5557684
BC112061.1 - 2 125 8327529
BG701467.1 - 3 518
BM768825.1 - 4 415
CB160001.1 - 4 366
CN347371.1 - 3 280
CR457005.1 - 2 81

View Full Product Details