Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001015881.1 | NP_001015881.1 | 3 | 483 | |
NM_001318468.1 | NP_001305397.1 | 4 | 951 | ||
NM_001318470.1 | NP_001305399.1 | 4 | 937 | ||
NM_004089.3 | NP_004080.2 | 3 | 688 | ||
NM_198057.2 | NP_932174.1 | 3 | 1004 | ||
XM_005262099.1 | XP_005262156.1 | 5 | 965 | ||
XM_005262100.1 | XP_005262157.1 | 4 | 840 | ||
XM_005262102.1 | XP_005262159.1 | 4 | 833 | ||
XM_005262103.3 | XP_005262160.1 | 4 | 943 | ||
XM_011530884.1 | XP_011529186.1 | 4 | 1039 | ||
XM_017029335.1 | XP_016884824.1 | 4 | 775 | ||
GenBank mRNA | AF183393.1 | - | 3 | 674 | |
AF228339.1 | - | 3 | 678 | ||
AK092645.1 | - | 4 | 890 | ||
AK092669.1 | - | 3 | 976 | ||
AK127938.1 | - | 3 | 628 | ||
AK300535.1 | - | 4 | 815 | ||
AL110191.1 | - | 3 | 687 | ||
AY007119.1 | - | 3 | 587 | ||
BC018148.1 | - | 3 | 370 | ||
BC072446.1 | - | 3 | 682 | 6165765 | |
BX647854.1 | - | 4 | 937 | ||
CR533450.1 | - | 3 | 426 | ||
Z50781.1 | - | 3 | 402 |