Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002872.4 | NP_002863.1 | 3 | 283 | |
XM_006724286.3 | XP_006724349.1 | 3 | 243 | ||
GenBank mRNA | AF498965.1 | - | 3 | 132 | |
BC001485.2 | - | 3 | 255 | 3509104 | |
BI908751.1 | - | 3 | 284 | ||
BQ934070.1 | - | 3 | 251 | ||
BT006919.1 | - | 3 | 132 | ||
CR456555.1 | - | 3 | 165 | ||
M29871.1 | - | 3 | 132 |