Gene Transcripts

Gene Symbol
HIKESHI
Entrez Gene ID
51501
Gene Name
Hikeshi, heat shock protein nuclear import factor
Gene Aliases
C11orf73, HLD13, HSPC138, HSPC179, L7RN6, OPI10
Chromosome Location
Chr. 11: 86302211 - 86345943 on Build GRCh38
UniGene ID
Hs.283322
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001322404.1 NP_001309333.1 2 384
NM_001322407.1 NP_001309336.1 3 558
NM_001322409.1 NP_001309338.1 3 488
NM_016401.3 NP_057485.2 2 400
NR_136324.1 2 384
XM_017017914.1 XP_016873403.1 2 390
XM_017017915.1 XP_016873404.1 2 177
GenBank mRNA AF151082.1 - 1 44
AF161487.1 - 2 384
AK001447.1 - 2 371
AK223198.1 - 2 371
AK298763.1 - 2 360
BC001677.2 - 2 273 2820772
BC006476.2 - 2 273 2820772
BC015991.2 - 2 365 2820691
BC018080.1 - 2 334 4798958
BC021621.1 - 2 365 2820691
BX332668.2 - 2 237

View Full Product Details