Gene Transcripts

Gene Symbol
LIPT1
Entrez Gene ID
51601
Gene Name
lipoyltransferase 1
Gene Aliases
LIPT1D
Chromosome Location
Chr. 2: 99154955 - 99163157 on Build GRCh38
UniGene ID
Hs.719172
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001204830.1 NP_001191759.1 3 676
NM_015929.3 NP_057013.1 3 478
NM_145197.2 NP_660198.1 4 606
NM_145198.2 NP_660199.1 4 594
NM_145199.2 NP_660200.1 2 352
NR_037935.1 4 739
NR_037936.1 3 415
GenBank mRNA AB017566.1 - 3 478
BC007001.1 - 3 398 3961227
BC009772.1 - 3 419 4091817
BC070145.1 - 3 456 4671058
BG717409.1 - 3 655
BQ422048.1 - 3 387

View Full Product Details