Gene Transcripts

Gene Symbol
CMC2
Entrez Gene ID
56942
Gene Name
C-X9-C motif containing 2
Gene Aliases
2310061C15Rik, C16orf61, DC13
Chromosome Location
Chr. 16: 80975800 - 81007046 on Build GRCh38
UniGene ID
Hs.55028
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020188.3 NP_064573.1 3 336
XM_017023461.1 XP_016878950.1 4 524
XM_017023462.1 XP_016878951.1 4 539
XM_017023463.1 XP_016878952.1 3 464
XM_017023467.1 XP_016878956.1 5 419
XM_017023468.1 XP_016878957.1 4 329
XM_017023469.1 XP_016878958.1 4 403
XM_017023470.1 XP_016878959.1 4 327
GenBank mRNA AF201935.1 - 3 312
AK123193.1 - 2 1691
BC032631.1 - 3 340 5399193

View Full Product Details