Gene Transcripts

Gene Symbol
THNSL2
Entrez Gene ID
55258
Gene Name
threonine synthase like 2
Gene Aliases
SOFAT, THS2, TSH2
Chromosome Location
Chr. 2: 88170295 - 88186637 on Build GRCh38
UniGene ID
Hs.739080
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001244676.1 NP_001231605.1 4 734
NM_001244678.1 NP_001231607.1 2 304
NM_018271.4 NP_060741.3 3 572
XM_005264400.4 XP_005264457.1 4 684
XM_005264401.4 XP_005264458.1 4 653
XM_005264402.3 XP_005264459.1 3 2473
XM_005264403.4 XP_005264460.1 3 2473
XM_005264405.4 XP_005264462.1 3 2473
XM_006712041.3 XP_006712104.1 3 2473
XM_006712042.3 XP_006712105.1 3 2473
XM_006712043.2 XP_006712106.1 3 2473
XM_006712044.3 XP_006712107.1 3 2473
XM_011532953.2 XP_011531255.1 3 2473
XM_011532954.2 XP_011531256.1 2 2278
XM_011532955.2 XP_011531257.1 3 418
GenBank mRNA AK001778.1 - 3 375
AK095303.1 - 3 2252
AK289486.1 - 3 404
AK315831.1 - 2 230
AL832746.1 - 1 104
BC035315.1 - 3 403
BC047758.2 - 1 143
BC064423.1 - 4 631 5752279
BI765911.1 - 3 486
HM185274.1 - 1 86

View Full Product Details