Gene Transcripts

Gene Symbol
RBP5
Entrez Gene ID
83758
Gene Name
retinol binding protein 5
Gene Aliases
CRBP-III, CRBP3, CRBPIII, HRBPiso
Chromosome Location
Chr. 12: 7119106 - 7130272 on Build GRCh38
UniGene ID
Hs.246046
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017020002.1 XP_016875491.1 4 1081
GenBank mRNA AF212239.1 - 4 693
AK223378.1 - 4 708
AY007436.1 - 4 689
BC029355.1 - 4 710 4613026

View Full Product Details