Gene Transcripts

Gene Symbol
PFDN4
Entrez Gene ID
5203
Gene Name
prefoldin subunit 4
Gene Aliases
C1, PFD4
Chromosome Location
Chr. 20: 54207963 - 54219953 on Build GRCh38
UniGene ID
Hs.91161
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002623.3 NP_002614.2 2 212
XM_017027879.1 XP_016883368.1 2 496
GenBank mRNA AK312059.1 - 2 88
BC010953.1 - 2 92 4273238
CN482830.1 - 2 212
U41816.1 - 2 73

View Full Product Details