Gene Transcripts

Gene Symbol
Tf
Entrez Gene ID
24825
Gene Name
transferrin
Gene Aliases
Tfn, Trf
Chromosome Location
Chr. 8: 111694570 - 111721275 on Build Rnor_6.0
UniGene ID
Rn.91296
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013110.1 NP_001013128.1 2, 3 246
GenBank mRNA AF476964.1 - 2, 3 277
AY327504.1 - 2, 3 215
BC087021.1 - 2, 3 246 7132382
D38380.1 - 2, 3 245
X77158.1 - 2, 3 232

Gene Symbol
Srprb
Entrez Gene ID
300965
Gene Name
signal recognition particle receptor, B subunit
Gene Aliases
Ab2-417, Ba1-667, Cc1-8
Chromosome Location
Chr. 8: 111673479 - 111687426 on Build Rnor_6.0
UniGene ID
Rn.138078
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AY325214.1 - 255
AY325230.1 - 255
AY325241.1 - 255

View Full Product Details