Gene Transcripts

Gene Symbol
Gm17689
Entrez Gene ID
100312948
Gene Name
predicted gene, 17689
Gene Aliases
Pate-g
Chromosome Location
Chr. 9: 36581278 - 36582635 on Build GRCm38
UniGene ID
Mm.475849
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001167584.1 NP_001161056.1 3 237
GenBank mRNA EU703627.1 - 3 237

View Full Product Details