Gene Transcripts

Gene Symbol
Map4k3
Entrez Gene ID
225028
Gene Name
mitogen-activated protein kinase kinase kinase kinase 3
Gene Aliases
4833416M01Rik, 4833416M07Rik, 9530052P13Rik, Glk, MAPKKKK3, MEKKK 3, MEKKK3, RAB8IPL1
Chromosome Location
Chr. 17: 80580513 - 80728205 on Build GRCm38
UniGene ID
Mm.311948
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290345.1 NP_001277274.1 15 1221
GenBank mRNA BQ442103.1 - 8 637
CF167620.1 - 4 289

View Full Product Details