Gene Transcripts

Gene Symbol
Prrc2c
Entrez Gene ID
226562
Gene Name
proline-rich coiled-coil 2C
Gene Aliases
1810043M20Rik, 9630039I18Rik, A630006J20, Bat2d, Bat2d1, Bat2l2, E130112L15Rik, Prrc3, mKIAA1096
Chromosome Location
Chr. 1: 162670727 - 162740564 on Build GRCm38
UniGene ID
Mm.245446
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081290.1 NP_001074759.1 16 4160
GenBank mRNA BC050871.1 - 1 22

View Full Product Details