Gene Transcripts

Gene Symbol
Mettl11b
Entrez Gene ID
240879
Gene Name
methyltransferase like 11B
Gene Aliases
Gm207, NTM1B
Chromosome Location
Chr. 1: 163701870 - 163725232 on Build GRCm38
UniGene ID
Mm.484621
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001143956.1 NP_001137428.1 3 551
GenBank mRNA BC157906.1 - 3 551
BC158115.1 - 3 528 9088188
BC172048.1 - 3 551 9054334

View Full Product Details