Gene Transcripts

Gene Symbol
Wdr76
Entrez Gene ID
241627
Gene Name
WD repeat domain 76
Gene Aliases
5830411K18Rik, 6030457D09, AA517836
Chromosome Location
Chr. 2: 121506723 - 121544860 on Build GRCm38
UniGene ID
Mm.293284
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290986.1 NP_001277915.1 11 1536
NM_001290987.1 NP_001277916.1 10 1421
NM_030234.2 NP_084510.2 11 1551
GenBank mRNA AK031592.1 - 11 1477
AK162181.1 - 11 1504
BC094676.1 - 10 1187 30615673
BC117821.1 - 10 1360
BC117822.1 - 11 1490 40092839

View Full Product Details