Gene Transcripts

Gene Symbol
Mtx3
Entrez Gene ID
382793
Gene Name
metaxin 3
Gene Aliases
4930470O13Rik, AA409304, AI853833, AU067765, EG624619, Gm1194, Gm6514
Chromosome Location
Chr. 13: 92844743 - 92858230 on Build GRCm38
UniGene ID
Mm.296770
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001162945.1 NP_001156417.1 9 2764

View Full Product Details