Gene Transcripts

Gene Symbol
Tpsb2
Entrez Gene ID
29268
Gene Name
tryptase beta 2
Gene Aliases
Mcpt6
Chromosome Location
Chr. 10: 14722672 - 14724464 on Build Rnor_6.0
UniGene ID
Rn.10183
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019180.2 NP_062053.2 2 120
GenBank mRNA D38455.1 - 3 130
U67909.1 - 2 121

View Full Product Details