Gene Transcripts

Gene Symbol
Rbm46
Entrez Gene ID
633285
Gene Name
RNA binding motif protein 46
Gene Aliases
EG633285, ENSMUSG00000033882, Gm761
Chromosome Location
Chr. 3: 82836521 - 82876533 on Build GRCm38
UniGene ID
Mm.265114
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001277170.1 NP_001264099.1 3 681

View Full Product Details