Gene Transcripts

Gene Symbol
Upf3a
Entrez Gene ID
67031
Gene Name
UPF3 regulator of nonsense transcripts homolog A (yeast)
Gene Aliases
2600001C03Rik, 4930546M19Rik, RENT3A, UPF3
Chromosome Location
Chr. 8: 13785215 - 13798538 on Build GRCm38
UniGene ID
Mm.44858
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025924.2 NP_080200.1 4, 5 587
GenBank mRNA AK011127.1 - 4, 5 564
AK016053.1 - 4, 5 550
AK033239.1 - 4, 5 568
AK145471.1 - 544
BC116988.1 - 4, 5 587 40126307
BC119035.1 - 4, 5 587 8733784

View Full Product Details