Gene Transcripts

Gene Symbol
Krtap4-6
Entrez Gene ID
68768
Gene Name
keratin associated protein 4-6
Gene Aliases
1110054P19Rik
Chromosome Location
Chr. 11: 99665043 - 99665960 on Build GRCm38
UniGene ID
Mm.416844
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026834.2 NP_081110.1 1 23
GenBank mRNA AK004258.1 - 1 24

View Full Product Details