Gene Transcripts

Gene Symbol
Isx
Entrez Gene ID
71597
Gene Name
intestine specific homeobox
Gene Aliases
9130012O13Rik, Pix-1
Chromosome Location
Chr. 8: 74873174 - 74893506 on Build GRCm38
UniGene ID
Mm.158735
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001294278.1 NP_001281207.1 4 1524
NM_027837.3 NP_082113.2 4 1565
GenBank mRNA AK018615.1 - 4 1566

View Full Product Details