Gene Transcripts
- Gene Symbol
- Ndufv2
- Entrez Gene ID
- 72900
- Gene Name
- NADH dehydrogenase (ubiquinone) flavoprotein 2
- Gene Aliases
- 2900010C23Rik
- Chromosome Location
- Chr. 17: 66078795 - 66101559 on Build GRCm38
- UniGene ID
- Mm.2206
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001278415.1 |
NP_001265344.1
|
3, 4 |
306 |
|
NM_028388.3 |
NP_082664.1
|
2, 3 |
209 |
|
GenBank mRNA |
AK013511.1 |
- |
2, 3 |
160 |
|
AK078351.1 |
- |
2, 3 |
153 |
|
AK146998.1 |
- |
2, 3 |
143 |
|
AK150404.1 |
- |
3, 4 |
243 |
|
AK151729.1 |
- |
3, 4 |
243 |
|
AK159460.1 |
- |
2, 3 |
126 |
|
AK166539.1 |
- |
2, 3 |
170 |
|
AK169143.1 |
- |
2, 3 |
180 |
|
BC030946.1 |
- |
2, 3 |
138 |
5003524 |
BY246389.1 |
- |
2, 3 |
210 |
|
CF106105.1 |
- |
1, 2 |
51 |
|
View Full Product Details